missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluorobenzene, 98+%

Product Code. 11494193
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494193 50 g 50g
11484193 10 g 10g
11404203 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494193 Supplier Thermo Scientific Alfa Aesar Supplier No. A10377.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 367-11-3 | C6H4F2 | 114.10 g/mol

1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-11-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000284
InChI Key GOYDNIKZWGIXJT-UHFFFAOYSA-N
Synonym o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene
PubChem CID 9706
ChEBI CHEBI:38583
IUPAC Name 1,2-difluorobenzene
SMILES FC1=CC=CC=C1F

Specifications

Melting Point -34°C
Density 1.171
Boiling Point 92°C to 94°C
Flash Point 1°C (33°F)
Odor Characteristic
Refractive Index 1.444
Quantity 50 g
UN Number UN1993
Beilstein 1905113
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 114.09
Percent Purity ≥98%
Chemical Name or Material 1,2-Difluorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Difluorobenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Difluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.