missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 97%

Product Code. 11490293
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490293 100 g 100g
11480293 25 g 25g
11400303 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490293 Supplier Thermo Scientific Alfa Aesar Supplier No. A11439.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 270573-71-2 | C6H6Na2O9S2 | 332.205 g/mol

1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, is used as a colorimetric reagent for iron, manganese, titanium, molybdenum, and complexes of zirconium. It is used as a metal indicator. It is used for Chelate titration, colorimetry, fluorometry and masking reagent. It is used in chemical research, pharmaceutical research and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, is used as a colorimetric reagent for iron, manganese, titanium, molybdenum, and complexes of zirconium. It is used as a metal indicator. It is used for Chelate titration, colorimetry, fluorometry and masking reagent. It is used in chemical research, pharmaceutical research and as a pharmaceutical intermediate.

Solubility
It is readily soluble in water.

Notes
Stable under normal conditions. Keep away from strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270573-71-2
Molecular Formula C6H6Na2O9S2
Molecular Weight (g/mol) 332.205
MDL Number MFCD00007473
InChI Key ZLRROLLKQDRDPI-UHFFFAOYSA-L
Synonym tiron monohydrate, sodium 4,5-dihydroxybenzene-1,3-disulfonate hydrate, unii-kht84q5m4r, 4,5-dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate, kht84q5m4r, disodium 4,5-dihydroxybenzene-1,3-disulfonate hydrate, 1,2-dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate, 1,3-benzenedisulfonicacid, 4,5-dihydroxy-, sodium salt, hydrate 1:2:1, 4,5-dihydroxybenzene-1,3-disulfonic acid, sodium salt, sodium salt, hydrate, sdd
PubChem CID 2723960
IUPAC Name disodium;4,5-dihydroxybenzene-1,3-disulfonate;hydrate
SMILES C1=C(C=C(C(=C1S(=O)(=O)[O-])O)O)S(=O)(=O)[O-].O.[Na+].[Na+]

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 4629740
Merck Index 14,9465
Solubility Information It is readily soluble in water.
Formula Weight 332.23 (314.21 Anhydrous)
Percent Purity 97%
Chemical Name or Material 1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate
compliance-icons
Product Identifier
  • 1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Dihydroxybenzene-3,5-disulfonic aciddisodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.