missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diisonicotinoylhydrazine, TRC

Product Code. 30006452
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006452 2.5 g 2.5g
30005577 1 g 1g
30011298 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006452 Supplier Toronto Research Chemicals Supplier No. TRCD4552752.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4329-75-3
Molecular Formula C12 H10 N4 O2
Molecular Weight (g/mol) 242.23
Synonym Isonicotinic Acid N'-(Pyridine-4-carbonyl)hydrazide, N,N'-Bis(4-picolinoyl)hydrazine, N,N'-Diisonicotinoylhydrazine, NSC 39301, NSC 52961
IUPAC Name N'-(pyridine-4-carbonyl)pyridine-4-carbohydrazide
SMILES O=C(NNC(=O)c1ccncc1)c2ccncc2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H10N4O2/c17-11(9-1-5-13-6-2-9)15-16-12(18)10-3-7-14-8-4-10/h1-8H,(H,15,17)(H,16,18)
Formula Weight 242.0804
Percent Purity >95%
Chemical Name or Material 1,2-Diisonicotinoylhydrazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.