missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxy-4,5-dinitrobenzene, 96%

Product Code. 11352577
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352577 1 g 1g
11362577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352577 Supplier Thermo Scientific Alfa Aesar Supplier No. L11706.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3395-03-7 | C8H8N2O6 | 228.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3395-03-7
MDL Number MFCD00082695

Specifications

Melting Point 131°C to 134°C
Molecular Formula C8H8N2O6
Quantity 1 g
UN Number UN2811
Beilstein 2055812
Synonym 4,5-dinitro veratrole, 1,2-dimethoxy-4,5-dinitro-benzene, 4,5-dimethoxy-1,2-dinitrobenzene, 4,5-dinitroveratrole, acmc-1ckho, ghl.pd_mitscher_leg0.933, benzene,1,2-dimethoxy-4,5-dinitro, benzene, 1,2-dimethoxy-4,5-dinitro
InChI Key WFDHPWTYKOAFBJ-UHFFFAOYSA-N
SMILES COC1=C(C=C(C(=C1)[N+](=O)[O-])[N+](=O)[O-])OC
IUPAC Name 1,2-dimethoxy-4,5-dinitrobenzene
Molecular Weight (g/mol) 228.16
PubChem CID 222989
Formula Weight 228.16
Percent Purity 96%
Chemical Name or Material 1,2-Dimethoxy-4,5-dinitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.