missing translation for 'onlineSavingsMsg'
Learn More

(1,2-Dioxoethylene)bis(iminoethylene) bis(3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate), TRC

Product Code. 30134299
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134299 2.5 g 2.5g
30133083 500 mg 500mg
30008576 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134299 Supplier Toronto Research Chemicals Supplier No. TRCD4875382.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70331-94-1
Molecular Formula C40 H60 N2 O8
Molecular Weight (g/mol) 696.91
Synonym Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, (1,2-dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl) ester (9CI), (oxalylbis(azanediyl))bis(ethane-2,1-diyl) bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate), 2,2'-Oxamidobis[ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 2,2'-Oxamidodiethyl bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], MD 697, MD 697 (antioxidant), N,N'-Bis(2-hydroxyethyl)oxamide bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Naugard XL 1, Naugard XL-I, Omnistab AN 697, ST 697, XL 1
IUPAC Name 2-[[2-[2-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxy]ethylamino]-2-oxoacetyl]amino]ethyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
SMILES CC(C)(C)c1cc(CCC(=O)OCCNC(=O)C(=O)NCCOC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C40H60N2O8/c1-37(2,3)27-21-25(22-28(33(27)45)38(4,5)6)13-15-31(43)49-19-17-41-35(47)36(48)42-18-20-50-32(44)16-14-26-23-29(39(7,8)9)34(46)30(24-26)40(10,11)12/h21-24,45-46H,13-20H2,1-12H3,(H,41,47)(H,42,48)
Formula Weight 696.435
Chemical Name or Material 2,2'-Oxamidobis[ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.