missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanedithiol, 98+%

Product Code. 10763904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763904 1 L Glass bottle 1 L
10738304 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10763904 Supplier Thermo Scientific Acros Supplier No. 117850010
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 540-63-6 | C2H6S2 | 94.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-63-6
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00004892
InChI Key VYMPLPIFKRHAAC-UHFFFAOYSA-N
Synonym 1,2-ethanedithiol, 1,2-dimercaptoethane, dithioglycol, ethylene mercaptan, ethylenedithiol, 1,2-ethanethiol, ethylenedimercaptan, dithioethyleneglycol, ethylene dithioglycol, s-ethylene dimercaptan
PubChem CID 10902
IUPAC Name ethane-1,2-dithiol
SMILES C(CS)S

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -41.0°C
Density 1.1230g/mL
Boiling Point 144.0°C to 146.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HSCH2CH2SH
Refractive Index 1.558
Quantity 1 L
Beilstein 01, 471
Fieser 01,356; 02,189; 05,290
Merck Index 15, 3780
Specific Gravity 1.123
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol
Formula Weight 94.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1, 2-Ethanedithiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.