missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)ethanol, 95%, Thermo Scientific Chemicals

Product Code. 15424557
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424557 10 g 10g
15434557 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424557 Supplier Thermo Scientific Alfa Aesar Supplier No. H31900.09
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Immiscible in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-26-1
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD00014401
InChI Key SXFYVXSOEBCFLV-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl ethanol, 2-fluorophenylmethylcarbinol, 2-fluoro-alpha-methylbenzyl alcohol, 1-2-fluorophenyl ethan-1-ol, o-fluorophenylmethylcarbinol, 2-fluorophenyl methyl carbinol, 2-fluorophenylethanol, 1-m-fluorophenyl ethanol, 1-2'-fluorophenyl ethanol, fluorophenylmethylcarbinol
PubChem CID 123066
IUPAC Name 1-(2-fluorophenyl)ethanol
SMILES CC(C1=CC=CC=C1F)O

Specifications

Quantity 10 g
Solubility Information Immiscible in water.
Formula Weight 140.16
Percent Purity 95%
Chemical Name or Material 1-(2-Fluorophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.