missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)ethanol, 95%, Thermo Scientific Chemicals

Product Code. 15434557
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15434557

Brand: Thermo Scientific Alfa Aesar H31900.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Immiscible in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-26-1
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD00014401
InChI Key SXFYVXSOEBCFLV-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl ethanol, 2-fluorophenylmethylcarbinol, 2-fluoro-alpha-methylbenzyl alcohol, 1-2-fluorophenyl ethan-1-ol, o-fluorophenylmethylcarbinol, 2-fluorophenyl methyl carbinol, 2-fluorophenylethanol, 1-m-fluorophenyl ethanol, 1-2'-fluorophenyl ethanol, fluorophenylmethylcarbinol
PubChem CID 123066
IUPAC Name 1-(2-fluorophenyl)ethanol
SMILES CC(C1=CC=CC=C1F)O

Specifications

Quantity 50 g
Solubility Information Immiscible in water.
Formula Weight 140.16
Percent Purity 95%
Chemical Name or Material 1-(2-Fluorophenyl)ethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.