missing translation for 'onlineSavingsMsg'
Learn More

1-(2-hydroxy-1-naphthyl)ethan-1-one, Thermo Scientific™

Product Code. 10279984
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10279984 5 g 5g
10416383 10 g 10g
10586134 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10279984 Supplier Thermo Scientific Maybridge Supplier No. SB01921DE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 574-19-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
InChI Key VUIOUIWZVKVFCI-UHFFFAOYSA-N
Synonym 2'-hydroxy-1'-acetonaphthone, 1-acetyl-2-naphthol, 1-2-hydroxynaphthalen-1-yl ethanone, 1-2-hydroxy-1-naphthalenyl ethanone, 1-2-hydroxy-1-naphthyl ethan-1-one, 2-hydroxy-1-acetonaphthone, 1-2-hydroxy-1-naphthyl ethanone, ethanone, 1-2-hydroxy-1-naphthalenyl, 1-acetyl-2-hydroxynaphthalene, 1'-acetonaphthone, 2'-hydroxy
PubChem CID 68455
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)ethanone
SMILES CC(=O)C1=C(C=CC2=CC=CC=C21)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 1-(2-Hydroxy-1-naphthyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.