missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Iodobenzyl)-1H-1,2,4-triazole, 97%, Thermo Scientific™

Product Code. 10347064
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347064 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10347064 Supplier Thermo Scientific Maybridge Supplier No. CC47511DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 876316-31-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H8IN3
MDL Number MFCD08741431
Quantity 1 g
Synonym 1-2-iodobenzyl-1h-1,2,4-triazole, 1-2-iodophenyl methyl-1,2,4-triazole, 1-2-iodophenyl methyl-1h-1,2,4-triazole, 1h-1,2,4-triazole,1-2-iodophenyl methyl, 1-2-iodobenzyl-1h-1,2,4 triazole
InChI Key NDJIYFFWZVACMJ-UHFFFAOYSA-N
SMILES IC1=CC=CC=C1CN1C=NC=N1
IUPAC Name 1-[(2-iodophenyl)methyl]-1,2,4-triazole
Molecular Weight (g/mol) 285.09
PubChem CID 18525817
Formula Weight 285.08
Percent Purity 97%
Chemical Name or Material 1-(2-Iodobenzyl)-1H-1, 2, 4-triazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.