missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(morpholin-4-ylmethyl)phenyl]methanamine, 97%, Thermo Scientific™

Product Code. 10719605
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10719605 10 g Amber Glass Bottle 10g
10163134 250 mg Amber Glass Bottle 250mg
10399602 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10719605 Supplier Thermo Scientific Maybridge Supplier No. CC48613EA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 91271-82-8
Molecular Formula C12H18N2O
Molecular Weight (g/mol) 206.289
InChI Key NMFAEZHWSZZJOA-UHFFFAOYSA-N
Synonym 2-morpholin-4-ylmethylbenzylamine, 1-2-morpholin-4-ylmethyl phenyl methanamine, 2-morpholin-4-ylmethyl-benzylamine, 1-2-morpholin-4-ylmethyl phenyl methylamine, 2-morpholinomethyl phenyl methanamine, 2-morpholin-4-ylmethyl phenyl methanamine, 2-morpholin-4-ylmethyl phenyl methylamine, 2-morpholin-4-ylmethylphenylamine, 2-morpholin-4-ylmethyl benzyl amine
PubChem CID 4962390
IUPAC Name [2-(morpholin-4-ylmethyl)phenyl]methanamine
SMILES C1COCCN1CC2=CC=CC=C2CN

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 206.29
Percent Purity 95%
Chemical Name or Material 1-[2-(Morpholin-4-ylmethyl)phenyl]methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.