missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediacetonitrile, 98%

Product Code. 11464514
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464514 5 g 5g
11474514 25 g 25g
11418710 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464514 Supplier Thermo Scientific Alfa Aesar Supplier No. A13142.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 613-73-0 | C10H8N2 | 156.19 g/mol

1,2-Phenylenediacetonitrile is used as API and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Phenylenediacetonitrile is used as API and intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-73-0
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00001905
InChI Key FWPFXBANOKKNBR-UHFFFAOYSA-N
Synonym 1,2-phenylenediacetonitrile, o-phenylenediacetonitrile, 1,2-bis cyanomethyl benzene, o-xylylene dicyanide, 2-2-cyanomethyl phenyl acetonitrile, o-benzenediacetonitrile, 2,2'-1,2-phenylene diacetonitrile, o-bis cyanomethyl benzene, 1,2-benzenediacetonitrile, o-benzendiacetonitrile
PubChem CID 69180
IUPAC Name 2-[2-(cyanomethyl)phenyl]acetonitrile
SMILES N#CCC1=CC=CC=C1CC#N

Specifications

Melting Point 57°C to 61°C
Quantity 5 g
UN Number UN3439
Beilstein 510075
Solubility Information Insoluble in water.
Formula Weight 156.19
Percent Purity 98%
Chemical Name or Material 1,2-Phenylenediacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.