missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridyl)piperazine, 98%

Product Code. 10478730 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10478730 25 g Glass bottle 25g
10383522 5 g Glass bottle 5g
10328893 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10478730 Supplier Thermo Scientific Acros Supplier No. 165900250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34803-66-2 | C9H13N3 | 163.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34803-66-2
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.22
InChI Key GZRKXKUVVPSREJ-UHFFFAOYSA-N
Synonym 1-2-pyridyl piperazine, 1-pyridin-2-yl piperazine, 1-pyridin-2-yl-piperazine, 1-2-pyridinyl piperazine, piperazine, 1-2-pyridinyl, 2-pyridylpiperazine, pyridinylpiperazine, 2-1-piperazinyl pyridine
PubChem CID 94459
IUPAC Name 1-pyridin-2-ylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=N2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 120°C to 122°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5930 to 1.595
Quantity 25 g
Specific Gravity 1.07
Solubility Information Solubility in water: >500g/L (20 c). Other solubilities: >50g/100mL methanol- >50g/100mL ethanol (20 c),>50g/100mL 2-propanol- >50g/100mL aceton (20 c),>50g/100mL n,n-dimethylformamide (20 c),>50g/100mL methylen chloride
Formula Weight 163.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(2-Pyridyl)piperazine
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Pyridyl)piperazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.