missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thien-2-ylphenyl)methanamine hydrochloride, ≥95%, Thermo Scientific™

Product Code. 10692035
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10692035 250 mg Amber Glass Bottle 250mg
10219332 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10692035 Supplier Thermo Scientific Maybridge Supplier No. CC35563CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 863991-95-1
Molecular Formula C11H12ClNS
Molecular Weight (g/mol) 225.73
MDL Number MFCD07772821
InChI Key XLJIHGHEJAGJJC-UHFFFAOYSA-N
Synonym 1-2-thien-2-ylphenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methylamine hydrochloride, 2-thiophen-2-yl phenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methanaminehydrochloride, 1-2-thiophen-2-yl phenyl methanamine hydrochloride, c11h11ns.hcl, 2-thiophen-2-yl-benzylamine hydrochloride, 2-2-thienyl phenyl methylamine, chloride, 2-thien-2-yl phenyl methylamine hydrochloride
PubChem CID 18525724
IUPAC Name 1-[2-(thiophen-2-yl)phenyl]methanamine hydrochloride
SMILES Cl.NCC1=CC=CC=C1C1=CC=CS1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 225.74
Percent Purity 95%
Chemical Name or Material 1-(2-Thien-2-ylphenyl)methanamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.