missing translation for 'onlineSavingsMsg'
Learn More

1-[(3,3-Diphenylpropyl)methylamino]-2-methyl-2-propanol, TRC

Product Code. 30135085
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135085 10 g 10g
17982885 1 g 1g
17922985 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135085 Supplier Toronto Research Chemicals Supplier No. TRCD49184010G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100442-33-9
Molecular Formula C20 H27 N O
Molecular Weight (g/mol) 297.43
Synonym 2-Propanol, 1-[(3,3-diphenylpropyl)methylamino]-2-methyl-, 1-[(3,3-Diphenylpropyl)methylamino]-2-methyl-2-propanol, 2-Methyl-1-[N-methyl-N-(3,3-diphenylpropyl)amino]propan-2-ol
IUPAC Name 1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-ol
SMILES CN(CCC(c1ccccc1)c2ccccc2)CC(C)(C)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H27NO/c1-20(2,22)16-21(3)15-14-19(17-10-6-4-7-11-17)18-12-8-5-9-13-18/h4-13,19,22H,14-16H2,1-3H3
Formula Weight 297.2093
Percent Purity >95%
Chemical Name or Material 1-[(3,3-Diphenylpropyl)methylamino]-2-methyl-2-propanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.