missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-tert-butylbenzene, 97+%

Product Code. 11339525
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11339525

Brand: Thermo Scientific Alfa Aesar L18832.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1460-02-2 | C18H30 | 246.438 g/mol

1,3,5-Tri-tert-butylbenzene is used in the preparation of sandwich complexes of scandium, yttrium and lanthanide ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tri-tert-butylbenzene is used in the preparation of sandwich complexes of scandium, yttrium and lanthanide ions.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1460-02-2
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008831
InChI Key GUFMBISUSZUUCB-UHFFFAOYSA-N
Synonym 1,3,5-tri-tert-butylbenzene, benzene, 1,3,5-tris 1,1-dimethylethyl, 1,3,5-tri-t-butylbenzene, benzene, 1,3,5-tri-tert-butyl, 1,3,5-tris tert-butyl benzene, pubchem13719, intermediates-zcf02115, acmc-209cw6, 2,4,6-tri-tert-butylphenyl, ksc490m7l
PubChem CID 15089
IUPAC Name 1,3,5-tritert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 69°C to 73°C
Boiling Point 121°C to 122°C (12 mmHg)
Quantity 50 g
Beilstein 1909579
Solubility Information Insoluble in water.
Formula Weight 246.44
Percent Purity ≥97%
Chemical Name or Material 1,3,5-Tri-tert-butylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.