missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%

Product Code. 15445907
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445907 1 g 1g
15455907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445907 Supplier Thermo Scientific Alfa Aesar Supplier No. H33116.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 84322-56-5 | C6F3I3 | 509.775 g/mol

1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84322-56-5
Molecular Formula C6F3I3
Molecular Weight (g/mol) 509.775
MDL Number MFCD06248899
InChI Key NTAZOPPTLZSXQH-UHFFFAOYSA-N
Synonym 1,3,5-trifluoro-2,4,6-triiodo-benzene, 1,3,5-trilfluoro-2,4,6-triiodobenzene, benzene,1,3,5-trifluoro-2,4,6-triiodo, pubchem10637, 1,3,5-trifluoro-2,4, 1,3,5-triiodotrifluorobenzene, 1,3,5-trifluoro-2,4,6-triiodob, benzene, 1,3,5-trifluoro-2,4,6-triiodo
PubChem CID 3852430
IUPAC Name 1,3,5-trifluoro-2,4,6-triiodobenzene
SMILES C1(=C(C(=C(C(=C1I)F)I)F)I)F

Specifications

Melting Point 152°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 509.77
Percent Purity 97%
Chemical Name or Material 1,3,5-Trifluoro-2,4,6-triiodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.