missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethoxy-2-nitrobenzene, 98%

Product Code. 11468703
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468703 5 g 5g
11478703 25 g 25g
11488703 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468703 Supplier Thermo Scientific Alfa Aesar Supplier No. A10473.06
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 14227-18-0 | C9H11NO5 | 213.189 g/mol

2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

Solubility
Soluble in water.

Notes
Desiccate at room temperature. Incompatabile to strong oxidizing agents, strong acids and bases. Store in a cool, dry, well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14227-18-0
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00016992
InChI Key VWYAWLZEMLQGJH-UHFFFAOYSA-N
PubChem CID 518903
IUPAC Name 1,3,5-trimethoxy-2-nitrobenzene
SMILES COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC

Specifications

Melting Point 150°C to 153°C
Quantity 5 g
Beilstein 2277262
Synonym 2,4,6-trimethoxynitrobenzene, 1-nitro-2,4,6-trimethoxybenzene, benzene,1,3,5-trimethoxy-2-nitro, benzene, 1,3,5-trimethoxy-2-nitro, acmc-20ap5l, maybridge1_001624, 1,3,5-trimethoxy-2-nitro-benzene, #
Solubility Information Soluble in water.
Formula Weight 213.19
Percent Purity 98%
Chemical Name or Material 1,3,5-Trimethoxy-2-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.