missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethoxybenzene, 99%

Product Code. 10746233 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10746233 10 g Glass bottle 10g
10562371 50 g Glass bottle 50g
10165733 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10746233 Supplier Thermo Scientific Acros Supplier No. 160550100
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 621-23-8 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-23-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00008385
InChI Key LKUDPHPHKOZXCD-UHFFFAOYSA-N
Synonym phloroglucinol trimethyl ether, sym-trimethoxybenzene, benzene, 1,3,5-trimethoxy, 1,3,5-trimethyoxybenzene, unii-00vji3vg3d, 1,3,5-trimethoxy benzene, o,o,o-1,3,5-trimethylresorcinol, 00vji3vg3d, dsstox_cid_25963, dsstox_rid_81259
PubChem CID 69301
ChEBI CHEBI:31038
IUPAC Name 1,3,5-trimethoxybenzene
SMILES COC1=CC(OC)=CC(OC)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 53.0°C
Color White to Cream
Density 85g/mL
Boiling Point 255.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(OCH3)3
Quantity 10 g
Beilstein 06, 1101
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents,soluble in methanol
Formula Weight 168.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3, 5-Trimethoxybenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1,3,5-Trimethoxybenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.