missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(4-aminophenoxy)benzene, TRC

Product Code. 30455880
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455880 500 mg -
30451731 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455880 Supplier Toronto Research Chemicals Supplier No. TRCT875005500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102852-92-6
Molecular Formula C24H21N3O3
Molecular Weight (g/mol) 399.44
Synonym 4,4',4' '-[1,3,5-Benzenetriyltris(oxy)]trisbenzenamine, 1,3,5-Tris(p-aminophenoxy)benzene, 135TAPOB
IUPAC Name 4-[3,5-bis(4-aminophenoxy)phenoxy]aniline
SMILES Nc1ccc(Oc2cc(Oc3ccc(N)cc3)cc(Oc4ccc(N)cc4)c2)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C24H21N3O3/c25-16-1-7-19(8-2-16)28-22-13-23(29-20-9-3-17(26)4-10-20)15-24(14-22)30-21-11-5-18(27)6-12-21/h1-15H,25-27H2
Formula Weight 399.1583
Chemical Name or Material 1,3,5-Tris(4-aminophenoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.