missing translation for 'onlineSavingsMsg'
Learn More

1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, 75%, mixture of isomers

Product Code. 10648001 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 10648001

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19437-42-4
Molecular Formula C10H5Na3O9S3
Molecular Weight (g/mol) 434.29
MDL Number MFCD00149263 MFCD00003989
InChI Key NJPKYOIXTSGVAN-UHFFFAOYSA-K
Synonym 1,3,6-naphthalenetrisulfonic acid, sodium salt, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:?
PubChem CID 73555608
IUPAC Name sodium;5,7-disulfonaphthalene-2-sulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=CC(=CC(=C2C=C1)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Beige or White to Yellow
Loss on Drying 8% max.
Packaging Glass bottle
Quantity 1 kg
Beilstein 11,229
Solubility Information Solubility in water: 600g/L (20°C)
Formula Weight 368.36
Percent Purity ≥90% (by difference)
Physical Form Powder
Chemical Name or Material 1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate
compliance-icons
Product Identifier
  • 1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, mixture of isomers
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.