missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-5-sulfonyl chloride, 95%, Thermo Scientific™

Product Code. 10442463
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10442463 250 mg Amber Glass Bottle 250mg
10656763 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442463 Supplier Thermo Scientific Maybridge Supplier No. CC01603CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 115010-10-1
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.623
InChI Key ICUBASIDCXDQAW-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxole-5-sulfonyl chloride, benzo 1,3 dioxole-5-sulfonyl chloride, 2h-1,3-benzodioxole-5-sulfonyl chloride, 1,3-benzodioxole-5-sulfonylchloride, 1,3-benzodioxole-5-sulphonyl chloride, acmc-20a4hu, 5-chlorosulphonyl-1,3-benzodioxole, benzo 1,3 dioxol-5-sulfonyl chloride, benzo 1,3 dioxol-5-sulphonyl chloride, benzo 1,3-dioxol-5-sulphonyl chloride
PubChem CID 4913401
IUPAC Name 1,3-benzodioxole-5-sulfonyl chloride
SMILES C1OC2=C(O1)C=C(C=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 220.63
Percent Purity 95%
Chemical Name or Material 1, 3-Benzodioxole-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.