missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-5-sulfonyl chloride, 95%

Product Code. 15400968
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400968 250 mg 250mg
15480958 1 g 1g
15490958 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400968 Supplier Thermo Scientific Alfa Aesar Supplier No. H54755.MD
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 115010-10-1 | C7H5ClO4S | 220.623 g/mol

1,3-Benzodioxole-5-sulfonyl chloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzodioxole-5-sulfonyl chloride is used as an organic chemical synthesis intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115010-10-1
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.623
MDL Number MFCD04974524
InChI Key ICUBASIDCXDQAW-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxole-5-sulfonyl chloride, benzo 1,3 dioxole-5-sulfonyl chloride, 2h-1,3-benzodioxole-5-sulfonyl chloride, 1,3-benzodioxole-5-sulfonylchloride, 1,3-benzodioxole-5-sulphonyl chloride, acmc-20a4hu, 5-chlorosulphonyl-1,3-benzodioxole, benzo 1,3 dioxol-5-sulfonyl chloride, benzo 1,3 dioxol-5-sulphonyl chloride, benzo 1,3-dioxol-5-sulphonyl chloride
PubChem CID 4913401
IUPAC Name 1,3-benzodioxole-5-sulfonyl chloride
SMILES C1OC2=C(O1)C=C(C=C2)S(=O)(=O)Cl

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 220.63
Percent Purity 97%
Chemical Name or Material 1,3-Benzodioxole-5-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.