missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea

Product Code. 16637052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16637052 1 g 1g
16647052 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16637052 Supplier Thermo Scientific Acros Supplier No. 465880010
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1060-92-0 | C17H8F12N2S | 500.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1060-92-0
Molecular Formula C17H8F12N2S
Molecular Weight (g/mol) 500.31
MDL Number MFCD00829878
InChI Key RWXWQJYJWJNJNW-UHFFFAOYSA-N
Synonym Schreiner's Thiourea Catalyst
IUPAC Name 1,3-bis[3,5-bis(trifluoromethyl)phenyl]thiourea
SMILES FC(F)(F)C1=CC(=CC(NC(=S)NC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)=C1)C(F)(F)F

Specifications

Alpha Vector BISBISTRIFLUOROMETHYLPHENYLTHIOUREA
Melting Point 150°C to 157°C
Color Brown to White
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
Formula Weight 500.3
Physical Form Crsytalline Powder or Crystals
Chemical Name or Material 1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
compliance-icons
Product Identifier
  • 1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.