missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[4-(4-pyridyl)-2-thiazolyl]benzene, 97%, Thermo Scientific™

Product Code. p-7039429
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454898 1 g 1g
15464898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454898 Supplier Thermo Scientific Alfa Aesar Supplier No. H58670.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The derivatives of this compound has been synthesized and then reacted with nitrate salts of copper, cadmium, and cobalt and copper hexafluororacetylacetonate to produce new one-dimensional and two-dimensional coordination polymers which display a variety of structural motifs.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421261-85-9
Molecular Formula C22H14N4S2
Molecular Weight (g/mol) 398.502
MDL Number MFCD22682999
InChI Key BAIVNAOKZPUEIV-UHFFFAOYSA-N
Synonym 1,3-bis 4-4-pyridyl-2-thiazolyl benzene, 4-2-3-4-pyridin-4-yl-1,3-thiazol-2-yl phenyl-1,3-thiazol-4-yl pyridine
PubChem CID 86277494
IUPAC Name 4-pyridin-4-yl-2-[3-(4-pyridin-4-yl-1,3-thiazol-2-yl)phenyl]-1,3-thiazole
SMILES C1=CC(=CC(=C1)C2=NC(=CS2)C3=CC=NC=C3)C4=NC(=CS4)C5=CC=NC=C5

Specifications

Melting Point 290°C to 296°C
Quantity 1 g
Formula Weight 398.49
Percent Purity 97%
Chemical Name or Material 1,3-Bis[4-(4-pyridyl)-2-thiazolyl]benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.