missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromobenzyl)-4-methylperhydro-1,4-diazepine, 97%, Thermo Scientific™

Product Code. 10174054
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10174054 1 g Amber Glass Bottle 1g
11366795 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174054 Supplier Thermo Scientific Maybridge Supplier No. CC55910DA
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 414885-80-6
Molecular Formula C13H19BrN2
Molecular Weight (g/mol) 283.21
MDL Number MFCD00811036
InChI Key XUIRKBKKWOVRIJ-UHFFFAOYSA-N
Synonym 1-3-bromobenzyl-4-methylperhydro-1,4-diazepine, 1-3-bromophenyl methyl-4-methyl-1,4-diazepane, 1-3-bromobenzyl-4-methylhomopiperazine, 1h-1,4-diazepine,1-3-bromophenyl methyl hexahydro-4-methyl, 1-3-bromobenzyl-4-methyl-1,4-diazepane, cambridge id 5266396
PubChem CID 764720
IUPAC Name 1-[(3-bromophenyl)methyl]-4-methyl-1,4-diazepane
SMILES CN1CCCN(CC2=CC(Br)=CC=C2)CC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 283.21
Percent Purity 97%
Chemical Name or Material 1-(3-Bromobenzyl)-4-methylperhydro-1, 4-diazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.