missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromophenyl)-1H-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10351543
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351543 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10351543 Supplier Thermo Scientific Maybridge Supplier No. CC32910DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 294877-33-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H7BrN2
MDL Number MFCD06797472
Quantity 1 g
Synonym 1-3-bromophenyl-1h-pyrazole, 1-3-bromophenyl pyrazole, 1-3-bromo-phenyl-1h-pyrazole, 3-1h-pyrazol-1-yl bromobenzene, 3-bromophenyl pyrazole, acmc-209h9b, 1h-pyrazole,1-3-bromophenyl, 1h-pyrazole, 1-3-bromophenyl
InChI Key KUWAIOZTKMAIPI-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Br)N2C=CC=N2
IUPAC Name 1-(3-bromophenyl)pyrazole
Molecular Weight (g/mol) 223.073
PubChem CID 7127804
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 1-(3-Bromophenyl)-1H-pyrazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.