missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane, 94%, Thermo Scientific™

Product Code. 11472240
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472240 1 g 1g
11482240 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472240 Supplier Thermo Scientific Alfa Aesar Supplier No. H29130.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18055-70-4
Molecular Formula C18H26OSi2
Molecular Weight (g/mol) 314.58
MDL Number MFCD09266163
InChI Key MEQYEVSZYLVDNX-UHFFFAOYSA-N
Synonym sym-tetramethyl-di-p-tolyl disiloxane, 1,3-di p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis p-tolyl-1,1,3,3-tetramethyl disiloxane, disiloxane, 1,1,3,3-tetramethyl-1,3-bis 4-methylphenyl, dimethyl 4-methylphenyl silyl oxy dimethyl 4-methylphenyl silane, amtsi070, 1,3-di-p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis 4-methylphenyl-1,1,3,3-tetramethyldisiloxane, 1,3-di 4-methylphenyl-1,1,3,3-tetramethyldisiloxane
PubChem CID 519510
IUPAC Name {[dimethyl(4-methylphenyl)silyl]oxy}dimethyl(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](C)(C)O[Si](C)(C)C1=CC=C(C)C=C1

Specifications

Quantity 1 g
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 314.57
Percent Purity 94%
Chemical Name or Material 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.