missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane, 94%, Thermo Scientific™

Product Code. 11482240
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Tamaño de la unidad
11482240 5 g 5g
11472240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11482240 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H29130.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 18055-70-4
Molecular Formula C18H26OSi2
Molecular Weight (g/mol) 314.58
MDL Number MFCD09266163
InChI Key MEQYEVSZYLVDNX-UHFFFAOYSA-N
Synonym sym-tetramethyl-di-p-tolyl disiloxane, 1,3-di p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis p-tolyl-1,1,3,3-tetramethyl disiloxane, disiloxane, 1,1,3,3-tetramethyl-1,3-bis 4-methylphenyl, dimethyl 4-methylphenyl silyl oxy dimethyl 4-methylphenyl silane, amtsi070, 1,3-di-p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis 4-methylphenyl-1,1,3,3-tetramethyldisiloxane, 1,3-di 4-methylphenyl-1,1,3,3-tetramethyldisiloxane
PubChem CID 519510
IUPAC Name {[dimethyl(4-methylphenyl)silyl]oxy}dimethyl(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](C)(C)O[Si](C)(C)C1=CC=C(C)C=C1

Especificaciones

Quantity 5 g
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 314.57
Percent Purity 94%
Chemical Name or Material 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.