missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butyl-2-methoxybenzene, 99%

Product Code. 15437787
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437787 250 g 250g
15417787 10 g 10g
15427787 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437787 Supplier Thermo Scientific Alfa Aesar Supplier No. H37487.30
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1516-95-6 | C15H24O | 220.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1516-95-6
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.36
MDL Number MFCD20483412
InChI Key OGIJRUWUWSZKAV-UHFFFAOYSA-N
Synonym 1,3-di-tert-butyl-2-methoxybenzene, benzene, 1,3-bis 1,1-dimethylethyl-2-methoxy, 2,6-di-tert-butylanisole
PubChem CID 12794694
IUPAC Name 1,3-ditert-butyl-2-methoxybenzene
SMILES COC1=C(C=CC=C1C(C)(C)C)C(C)(C)C

Specifications

Color Colorless to Yellow
Boiling Point 124°C (9 mmHg)
Quantity 250 g
Formula Weight 220.35
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,3-Di-tert-butyl-2-methoxybenzene

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.