missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibenzoylbenzene, 98+%

Product Code. 11495103
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495103 5 g 5g
11405113 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495103 Supplier Thermo Scientific Alfa Aesar Supplier No. A12768.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3770-82-9 | C20H14O2 | 286.33 g/mol

1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.

Solubility
Sparingly Soluble (4.0E-3 g/L) (25°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3770-82-9
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00014086
InChI Key MJQHDSIEDGPFAM-UHFFFAOYSA-N
Synonym 1,3-dibenzoylbenzene, 1,3-phenylenebis phenylmethanone, m-dibenzoylbenzene, methanone, 1,3-phenylenebis phenyl, 3-benzoylphenyl phenyl methanone, m-dibenzoyl benzene, methanone, 1,1'-1,3-phenylene bis 1-phenyl, phenyl 3-phenylcarbonyl phenyl ketone
PubChem CID 77388
IUPAC Name (3-benzoylphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 98°C to 99°C
Quantity 5 g
Beilstein 1973337
Solubility Information Sparingly Soluble (4.0E-3g/L) (25°C).
Formula Weight 286.33
Percent Purity ≥98%
Chemical Name or Material 1,3-Dibenzoylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.