missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-fluorobenzene, 98%, ACROS Organics™

Product Code. 10406622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10406622 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10406622 Supplier Thermo Scientific Acros Supplier No. 263460050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. Rückgaberichtlinie anzeigen

Chemische Identifikatoren

CAS 1435-51-4
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00061119
InChI Key ASWYHZXKFSLNLN-UHFFFAOYSA-N
Synonym 3,5-dibromofluorobenzene, 3,5-dibromo-1-fluorobenzene, 1,3-dibromo-5-fluoro-benzene, benzene, 1,3-dibromo-5-fluoro, pubchem1052, acmc-1c2vf, intermediates-zcf02096, 3,5-bromo-1-fluorophenyl, 1,3dibromo-5-fluorobenzene, 3,5-dibromo-fluoro-benzene
PubChem CID 137003
IUPAC Name 1,3-dibromo-5-fluorobenzene
SMILES C1=C(C=C(C=C1Br)Br)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 2.0180g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br2C6H3F
Refractive Index 1.5760 to 1.5780 (20°C, 589nm)
Quantity 5 g
Formula Weight 253.9
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1, 3-Dibromo-5-fluorobenzene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.