missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromobenzene, 97%

Product Code. 10710891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10710891 10 mL Glass bottle 10mL
10417390 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10710891 Leverandør Thermo Scientific Acros Leverandørnr. 186640100
 Restricted Item
Estimated Shipment: 12-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 108-36-1 | C6H4Br2 | 235.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 108-36-1
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000078
InChI Key JSRLURSZEMLAFO-UHFFFAOYSA-N
Synonym m-dibromobenzene, benzene, 1,3-dibromo, benzene, m-dibromo, unii-74ef6kh8tc, 1,3-dibromo-benzene, 74ef6kh8tc, 1,3-dibromo benzene, 3,5-dibromobenzene, 1,3-dibrormobenzene, 1, 3-dibromobenzene
PubChem CID 7927
ChEBI CHEBI:37151
IUPAC Name 1,3-dibromobenzene
SMILES BrC1=CC(Br)=CC=C1

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point -7°C
Color Colorless to Yellow
Density 1.9500g/mL
Boiling Point 218°C to 219°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H4Br2
Refractive Index 1.6070 to 1.609
Quantity 10 mL
Beilstein 05, 211
Specific Gravity 1.95
Solubility Information Solubility in water: insoluble
Formula Weight 235.91
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 3-Dibromobenzene, 97%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 1,3-Dibromobenzene
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.