missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromotetrafluorobenzene, 98+%

Product Code. 11453637
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453637 5 g 5g
11443637 1 g 1g
11463637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453637 Supplier Thermo Scientific Alfa Aesar Supplier No. A17392.06
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1559-87-1 | C6Br2F4 | 307.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1559-87-1
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.87
MDL Number MFCD00031533
InChI Key UCWKDDQEZQRGDR-UHFFFAOYSA-N
PubChem CID 73793
IUPAC Name 1,3-dibromo-2,4,5,6-tetrafluorobenzene
SMILES FC1=C(F)C(Br)=C(F)C(Br)=C1F

Specifications

Density 2.18
Boiling Point 85°C to 88°C (18 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.516
Quantity 5 g
Synonym 1,3-dibromotetrafluorobenzene, benzene, 1,3-dibromotetrafluoro, benzene, 1,3-dibromo-2,4,5,6-tetrafluoro, 1,3-dibromo-2,4,5,6-tetrafluoro-benzene, benzene,1,3-dibromo-2,4,5,6-tetrafluoro, acmc-1byn7, benzene,3-dibromotetrafluoro, 1,3-dibromotetrafluorobenzene, 98+%
Formula Weight 307.87
Percent Purity ≥98%
Chemical Name or Material 1,3-Dibromotetrafluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.