missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-fluorobenzene, 98%, Thermo Scientific™

Product Code. 11417094
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417094 5 g 5g
11434060 1 g 1g
11427094 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417094 Supplier Thermo Scientific Alfa Aesar Supplier No. B21526.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2268-05-5
MDL Number MFCD00010307

Specifications

Melting Point 36°C to 40°C
Boiling Point 168°C to 169°C
Flash Point 73°C (163°F)
Molecular Formula C6H3Cl2F
Quantity 5 g
Beilstein 1862513
Synonym 2,6-dichlorofluorobenzene, 1,3-dichloro-2-fluoro-benzene, 2,6-dichloro-1-fluorobenzene, 2,6-dichloro fluoro benzene, benzene,1,3-dichloro-2-fluoro, benzene, 1,3-dichloro-2-fluoro, pubchem2311, 2,6-dichlorfluorbenzol, acmc-2097ae, 2,6-dichloro fluorobenzene
InChI Key JORVCRLRRRRLFI-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)F)Cl
IUPAC Name 1,3-dichloro-2-fluorobenzene
Molecular Weight (g/mol) 164.99
PubChem CID 137520
Formula Weight 165
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-fluorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.