missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-4-fluorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11479086
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479086 50 g 50g
11469086 10 g 10g
11489086 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479086 Supplier Thermo Scientific Alfa Aesar Supplier No. A17953.18
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1435-48-9
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.988
MDL Number MFCD00000580
InChI Key BDJZCCWUSOZUQG-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-fluorobenzene, 2,4-dichlorofluorobenzene, benzene, 2,4-dichloro-1-fluoro, dcfb, benzene,2,4-dichloro-1-fluoro, 2,4-dichloro-1-fluoro-benzene, pubchem2312, acmc-209cqu, 2,4-dichlorofluobenzene, 1,3-dichlorofluorobenzene
PubChem CID 123112
IUPAC Name 2,4-dichloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)F

Specifications

Melting Point -23°C
Density 1.409
Boiling Point 172°C to 174°C
Flash Point 79°C (174°F)
Refractive Index 1.525
Quantity 50 g
UN Number UN2810
Beilstein 2500791
Formula Weight 165
Percent Purity 99%
Chemical Name or Material 1,3-Dichloro-4-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.