missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethyl-1,3-diphenylurea, TRC

Product Code. 17973615
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973615 1 g 1g
17930805 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17973615 Supplier Toronto Research Chemicals Supplier No. TRCD4728801G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-98-3
Molecular Formula C17 H20 N2 O
Molecular Weight (g/mol) 268.35
Synonym N,N'-Diethyl-N,N'-diphenylurea, Centralite 1, 1,3-Diethyl-1,3-diphenylurea, Carbamite, Centralite, Centralite I, Ethyl Centralite, N,N'-Diethyl-N,N'-diphenylurea, N,N'-Diethylcarbanilide, NSC 28779, NSC 44038, sym-Diethyldiphenylurea;
IUPAC Name 1,3-diethyl-1,3-diphenylurea
SMILES CCN(C(=O)N(CC)c1ccccc1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H20N2O/c1-3-18(15-11-7-5-8-12-15)17(20)19(4-2)16-13-9-6-10-14-16/h5-14H,3-4H2,1-2H3
Formula Weight 268.1576
Percent Purity >95%
Chemical Name or Material Centralit 1
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.