missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethyl-2-thiobarbituric acid, 99%

Product Code. p-3778260 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
5g
This item is not returnable. View return policy

Product Code. 10373712

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5217-47-0 | C8H12N2O2S | 200.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5217-47-0
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.26
MDL Number MFCD00006676
InChI Key SHBTUGJAKBRBBJ-UHFFFAOYSA-N
Synonym 1,3-diethyl-2-thiobarbituric acid, 1,3-diethylthiobarbituric acid, n,n'-diethylthiobarbituric acid, n,n'-diethyl-2-thiobarbituric acid, 1,3-diethyl-2-thioxodihydropyrimidine-4,6 1h,5h-dione, unii-bxb40m6a7w, 4,6 1h,5h-pyrimidinedione, 1,3-diethyldihydro-2-thioxo, barbituric acid, 1,3-diethyl-2-thio, bxb40m6a7w, 1,3-diethyldihydro-2-thioxopyrimidine-4,6 1h,5h-dione
PubChem CID 78890
IUPAC Name 1,3-diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione
SMILES CCN1C(=O)CC(=O)N(CC)C1=S

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 105°C to 109°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 24, IV, 1885
Formula Weight 200.25
Percent Purity ≥98.5%
Physical Form Crystals or Powder
Chemical Name or Material 1, 3-Diethyl-2-thiobarbituric acid
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Diethyl-2-thiobarbituric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.