missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethylbenzene, 97+%

Product Code. 11375627
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
10mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375627 2 mL 2mL
11365627 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375627 Supplier Thermo Scientific Alfa Aesar Supplier No. 043369.S0
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 141-93-5 | C10H14 | 134.222 g/mol

1,3-Diethylbenzene is used as an intermediate and solvent. It is also used in the preparation of divinyl benzene. Further, it is used in powderless etching.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-93-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009260
InChI Key AFZZYIJIWUTJFO-UHFFFAOYSA-N
Synonym m-diethylbenzene, benzene, 1,3-diethyl, benzene, m-diethyl, m-ethylethylbenzene, unii-zm2x7i1g8y, zm2x7i1g8y, ethylated benzene, benzene, ethylated, meta-diethylbenzene, polyethylated benzene
PubChem CID 8864
IUPAC Name 1,3-diethylbenzene
SMILES CCC1=CC(=CC=C1)CC

Specifications

Color Colorless
Density 0.86 g/mL
Boiling Point 182°C
Flash Point 55°C (131°F)
Assay Percent Range >97%
Odor Aromatic
Refractive Index 1.496
Quantity 2 mL
UN Number UN2049
Solubility Information Miscible with ethanol,benzene,carbon tetrachloride,ethyl ether and acetone. Immiscible with water and diethylbenzene.
Formula Weight 134.22
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1,3-Diethylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.