accessibility menu, dialog, popup

UserName

CH3CH3

CAS RN 141-93-5

IUPAC Name: 1,3-diethylbenzene
Synonyms: M-diethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: AFZZYIJIWUTJFO-UHFFFAOYSA-N
SMILES: CCC1=CC(CC)=CC=C1
Solvent or Matrix 
  • (2)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)