missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethoxybenzene, 98%

Product Code. 11415373
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415373 100 g 100g
11425373 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415373 Supplier Thermo Scientific Alfa Aesar Supplier No. A13380.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 151-10-0 | C8H10O2 | 138.17 g/mol

1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

Solubility
Miscible with toluene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-10-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008384
InChI Key DPZNOMCNRMUKPS-UHFFFAOYSA-N
Synonym m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
PubChem CID 9025
IUPAC Name 1,3-dimethoxybenzene
SMILES COC1=CC(OC)=CC=C1

Specifications

Melting Point -52°C
Density 1.065
Boiling Point 217°C to 218°C
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.525
Quantity 100 g
Beilstein 878582
Solubility Information Miscible with toluene.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,3-Dimethoxybenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.