missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF, AcroSeal™

Product Code. 11985991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985991 100 mL AcroSeal™ Glass Bottle 100mL
11995991 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985991 Supplier Thermo Scientific Acros Supplier No. 438861000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 180675-22-3 | C4H7BrMgO2 | 191.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180675-22-3 , 109-99-9
Molecular Formula C4H7BrMgO2
Molecular Weight (g/mol) 191.31
MDL Number MFCD01321271
InChI Key ZDNTZRAFJZERSL-UHFFFAOYSA-M
Synonym 1,3-dioxolan-2-ylmethyl magnesium bromide solution, magnesium 2-methanidyl-1,3-dioxolane bromide, magnesium 2+ ion 1,3-dioxolan-2-ylmethanide bromide, grignard reagent
PubChem CID 4247254
IUPAC Name magnesium;2-methanidyl-1,3-dioxolane;bromide
SMILES Br[Mg]CC1OCCO1

Specifications

CAS Min % 8.14, 87.79
CAS Max % 12.21, 91.86
Color Brown to White
Density 0.9400g/mL
Boiling Point 65.0°C to 67.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.94
Formula Weight 191.31
Physical Form Powder
Chemical Name or Material (1, 3-Dioxolan-2-ylmethyl)magnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.