missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.8%, anhydrous, stabilized with 75 ppm BHT, AcroSeal™

Product Code. 12140910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12140910 1 L AcroSeal™ Glass Bottle 1 L
11986201 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12140910 Supplier Thermo Scientific Acros Supplier No. 431560010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 646-06-0 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0 , 128-37-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

CAS Min % 99.75
CAS Max % 100.0
Melting Point -95°C
Density 1.0600g/mL
Boiling Point 74.0°C to 75.0°C
Flash Point −6°C
Ignition Residue 0.0005% max
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3990 to 1.4010
Quantity 1 L
Residue after Evaporation 0.0005% max.
Specific Gravity 1.06
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and acetone
Stabilizer 75ppm (BHT)
Viscosity 0.66 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 99.8%
Physical Form Liquid
Chemical Name or Material 1, 3-Dioxolane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dioxolane, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.