missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Diphenoxy-2-propanol, 98+%

Product Code. 11474507
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474507 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474507 Supplier Alfa Aesar™ Supplier No. B20503.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 622-04-8
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00091115
InChI Key NKCVHIPHZCIEFC-UHFFFAOYSA-N
Synonym 1,3-diphenoxy-2-propanol, glycerol 1,3-diphenyl ether, 1,3-difenoxy-2-propanol, 2-propanol, 1,3-diphenoxy, unii-rto1if8e7z, 1,2,3-propanetriol, 1,3-diphenyl diether, glyceryl alpha,gamma-diphenyl ether, rto1if8e7z, 2-hydroxy-3-phenoxypropoxy benzene, glyceryl .alpha.,.gamma.-diphenyl ether
PubChem CID 12140
IUPAC Name 1,3-diphenoxypropan-2-ol
SMILES OC(COC1=CC=CC=C1)COC1=CC=CC=C1

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
Formula Weight 244.29
Percent Purity ≥98%
Chemical Name or Material 1,3-Diphenoxy-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.