missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylacetone, 98+%

Product Code. 11397855
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397855 100 g 100g
11387855 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397855 Supplier Thermo Scientific Alfa Aesar Supplier No. L02830.22
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102-04-5 | C15H14O | 210.28 g/mol

1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-04-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004795
InChI Key YFKBXYGUSOXJGS-UHFFFAOYSA-N
Synonym 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
PubChem CID 7593
IUPAC Name 1,3-diphenylpropan-2-one
SMILES O=C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 32°C to 36°C
Density 1.04
Boiling Point 330°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 974767
Solubility Information Slightly soluble in water.
Formula Weight 210.28
Percent Purity ≥98%
Chemical Name or Material 1,3-Diphenylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.