missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylpropane, 98%

Product Code. 11322746
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322746 2 g 2g
11332746 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322746 Supplier Thermo Scientific Alfa Aesar Supplier No. L08849.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1081-75-0 | C15H16 | 196.29 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1081-75-0
Molecular Formula C15H16
Molecular Weight (g/mol) 196.29
MDL Number MFCD00043574
InChI Key VEAFKIYNHVBNIP-UHFFFAOYSA-N
Synonym 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci
PubChem CID 14125
ChEBI CHEBI:34060
IUPAC Name 3-phenylpropylbenzene
SMILES C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Density 0.98
Boiling Point 158°C to 159°C (10 mmHg)
Linear Formula C6H5(CH2)3C6H5
Refractive Index 1.56
Quantity 2 g
Beilstein 2044541
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material 1,3-Diphenylpropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.