missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Iodobenzyl)-1H-1,2,4-triazole, ≥95%, Thermo Scientific™

Product Code. 10545174
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10545174 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10545174 Supplier Thermo Scientific Maybridge Supplier No. CC45411DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 615534-73-1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H8IN3
MDL Number MFCD08271930
Quantity 1 g
Synonym 1-3-iodobenzyl-1h-1,2,4-triazole, 1-3-iodophenyl methyl-1,2,4-triazole, 1h-1,2,4-triazole,1-3-iodophenyl methyl, 1-3-iodobenzyl-1h-1,2,4 triazole, 3-1h-1,2,4-triazol-1-ylmethyl iodobenzene, 1-3-iodophenyl methyl-1h-1,2,4-triazole
InChI Key RFHFJBONYNRFAA-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)I)CN2C=NC=N2
IUPAC Name 1-[(3-iodophenyl)methyl]-1,2,4-triazole
Molecular Weight (g/mol) 285.088
PubChem CID 18525776
Formula Weight 285.08
Percent Purity 95%
Chemical Name or Material 1-(3-Iodobenzyl)-1H-1, 2, 4-triazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.