missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Iodobenzyl)-1h-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10273053
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10273053 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10273053 Supplier Thermo Scientific Maybridge Supplier No. CC56111DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 884507-56-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9IN2
MDL Number MFCD08435919
Quantity 1 g
Synonym 1-3-iodobenzyl-1h-pyrazole, 1-3-iodophenyl methyl pyrazole, 1h-pyrazole,1-3-iodophenyl methyl, 1-3-iodophenyl methyl-1h-pyrazole
InChI Key GWLQKUUXEMKUEW-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)I)CN2C=CC=N2
IUPAC Name 1-[(3-iodophenyl)methyl]pyrazole
Molecular Weight (g/mol) 284.1
PubChem CID 18525879
Formula Weight 284.1
Percent Purity 97%
Chemical Name or Material 1-(3-Iodobenzyl)-1H-pyrazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.