missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methylphenyl)piperazine, 99%, Thermo Scientific™

Product Code. 10173011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10173011 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10173011 Supplier Thermo Scientific Acros Supplier No. 345950010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 41186-03-2
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00040734
InChI Key JIWHIRLNKIUYSM-UHFFFAOYSA-N
Synonym 1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine
PubChem CID 83111
IUPAC Name 1-(3-methylphenyl)piperazine
SMILES CC1=CC(=CC=C1)N2CCNCC2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5755 to 1.5785
Quantity 1 g
Formula Weight 176.26
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 1-(3-Methylphenyl)piperazine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(3-Methylphenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.