missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(tert-Butyl)phenyl]hydrazine hydrochloride 97%, Thermo Scientific™

Product Code. 10748835
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10748835 5 g 5g
10631365 250 mg 250mg
10376772 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10748835 Supplier Thermo Scientific Maybridge Supplier No. TL00869DE
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306937-27-9
Molecular Formula C10H17ClN2
Molecular Weight (g/mol) 200.71
InChI Key XKLKKAKHNGXDPA-UHFFFAOYSA-N
Synonym 1-3-tert-butyl phenyl hydrazine hydrochloride, 3-tert-butylphenyl hydrazine hydrochloride, 1-3-tert-butyl phenyl hydrazinehydrochloride, 1-tert-butyl-3-hydrazinobenzene, 3-tert-butyl phenylhydrazine, chloride, 3-tert-butylphenylhydrazine hydrochloride, 3-tert-butyl-phenylhydrazine hydrochloride, 3-tert-butyl phenyl hydrazine hydrochloride
PubChem CID 2801514
IUPAC Name (3-tert-butylphenyl)hydrazine;hydrochloride
SMILES CC(C)(C)C1=CC(=CC=C1)NN.Cl

Specifications

Packaging Amber glass bottle
Quantity 5 g
Formula Weight 200.71
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-[3-(tert-Butyl)phenyl]hydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.